Learn More
CELLTREAT Scientific Products 5mL Black Macro Centrifuge Tube, Non-sterile
SDP
Catalog No. 50969830
Encompass_Preferred
CELLTREAT Scientific Products 5mL Black Macro Centrifuge Tube. 5mL Macro Centrifuge Tubes offer the convenience of a micro centrifuge tube with the ability to process sample volumes up to 5mL. Tube diameter and shape is the same as a standard 15mL conical bottom centrifuge tube, enabling use with existing equipment. Tubes have frosted marking spot and engraved graduations. Attached flat top cap is easy to open and close with one hand. Center of the cap includes a thin """"membrane"""" area designed for easy puncture and access by a syringe/needle. Black tubes protect light sensitive compounds and eliminate the need to wrap tubes with aluminum foil. RNase/DNase free and non-pyrogenic. Max force (RCF) g 25000. Temperature range of -86¿C to 80¿C. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 100/re-sealable bag, 100/case
Catalog No. 50-969-830 Supplier CELLTREAT Scientific Products Supplier No. 229439
CELLTREAT Scientific Products 5mL Black Macro Centrifuge Tube. 5mL Macro Centrifuge Tubes offer the convenience of a micro centrifuge tube with the ability to process sample volumes up to 5mL. Tube diameter and shape is the same as a standard 15mL conical bottom centrifuge tube, enabling use with existing equipment. Tubes have frosted marking spot and engraved graduations. Attached flat top cap is easy to open and close with one hand. Center of the cap includes a thin """"membrane"""" area designed for easy puncture and access by a syringe/needle. Black tubes protect light sensitive compounds and eliminate the need to wrap tubes with aluminum foil. RNase/DNase free and non-pyrogenic. Max force (RCF) g 25000. Temperature range of -86¿C to 80¿C. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 100/re-sealable bag, 100/case