Learn More
Research Products International Corp 5mL Micro-Tube Rack
SDP
Catalog No. 501367746
Encompass
Economical rack for handling or transporting 5ml capacity Micro-Tubes and other 13-16mm diameter tubes. Rack features convenient carrying handles and a bottom tier for additional support. Compact design accommodates up to forty tubes in a 4 x 10 array. Made from autoclavable white polypropylene for durability and chemical resistance.
Catalog No. 50-136-7746 Supplier Research Products International Corp Supplier No. 249120
Economical rack for handling or transporting 5ml capacity Micro-Tubes and other 13-16mm diameter tubes. Rack features convenient carrying handles and a bottom tier for additional support. Compact design accommodates up to forty tubes in a 4 x 10 array. Made from autoclavable white polypropylene for durability and chemical resistance.