Learn More
Chemglass Life Sciences 5mL Micro Volumetric Flask, Class A, Tolerance: +-0.02mL
SDP
Catalog No. CG160404
Encompass_Preferred
5mL Micro Volumetric Flask, Class A, Tolerance: ¿0.02mL. Class ¿A¿ micro volumetric flask calibrated to contain. Flask has a GPI 13-425 top thread and is supplied complete with an open top screw cap and PTFE faced silicone septa. Flasks have hex base for stability and ¿V¿ shaped interior bottom for total sample retrieval. See MW-102 for replacement caps and MW-104 for replacement septa.
Catalog No. CG160404 Supplier Chemglass Life Sciences Supplier No. CG160404
5mL Micro Volumetric Flask, Class A, Tolerance: ¿0.02mL. Class ¿A¿ micro volumetric flask calibrated to contain. Flask has a GPI 13-425 top thread and is supplied complete with an open top screw cap and PTFE faced silicone septa. Flasks have hex base for stability and ¿V¿ shaped interior bottom for total sample retrieval. See MW-102 for replacement caps and MW-104 for replacement septa.