Learn More
Epoch Life Science Inc EconoSpin® Micro DNA/RNA Columns, 5ul Elution Volume w/ lid (250 columns with collection tubes)
SDP
Catalog No. NC0857585
Encompass_Preferred
Silica membrane micro spin columns with lid and collection tubes for gel PCR & plasmid purification. Sample volume: 850 ul. Binding capacity: 40 ug plasmid DNA. Minimum elution volume: 5 ul. Compatible with third party buffer sets. 18 month shelf life. Used by labs around the world.
Catalog No. NC0857585 Supplier Epoch Life Science Inc Supplier No. 3010250
Silica membrane micro spin columns with lid and collection tubes for gel PCR & plasmid purification. Sample volume: 850 ul. Binding capacity: 40 ug plasmid DNA. Minimum elution volume: 5 ul. Compatible with third party buffer sets. 18 month shelf life. Used by labs around the world.