Learn More
S2 Media 5mL Normal Saline, 0.85% (w/v) NaCl Solution, Sterile, 16x125mm Borosilicate Glass Tube, 50/box
Catalog No. 502995916
Encompass_Preferred
Normal Saline, 0.85% (w/v) NaCl Solution ready-to-use tubes are intended for use as a buffer solution, diluent, and in the preparation of bacterial suspensions. This product is not intended for growth promotion; therefore, growth performance testing with specified microorganisms is not indicated.• Normal Saline (0.85% NaCl) should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5916 Supplier S2 Media Supplier No. 5503
Normal Saline, 0.85% (w/v) NaCl Solution ready-to-use tubes are intended for use as a buffer solution, diluent, and in the preparation of bacterial suspensions. This product is not intended for growth promotion; therefore, growth performance testing with specified microorganisms is not indicated.• Normal Saline (0.85% NaCl) should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.