Learn More
Bellco Glass, Inc. 5mL Pipet, Individually
SDP
Catalog No. 502329763
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identification Pack Description50 Individually Wrapped/BagCase Quantity200SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorBluePrimary MaterialPolystyreneMaximum Volume5mLGraduatedYesGraduation Increments (Subdivisions)1/10mLRNase/DNase FreeYes
Catalog No. 50-232-9763 Supplier Bellco Glass, Inc. Supplier No. 229205B00
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identification Pack Description50 Individually Wrapped/BagCase Quantity200SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorBluePrimary MaterialPolystyreneMaximum Volume5mLGraduatedYesGraduation Increments (Subdivisions)1/10mLRNase/DNase FreeYes