Learn More
CELLTREAT Scientific Products 5mL Pipette Tips, Bulk Bag, Non-sterile
SDP
Catalog No. 5000144327
Encompass_Preferred
Sterile, accurate liquid transfer of larger volumes of media and solutionsUniversal fit is compatible with a range of popular pipettesMore precise than serological pipets for volumes of 1 to 10mLTips have molded graduations, with 0.5mL increments
Catalog No. 50-001-44327 Supplier CELLTREAT Scientific Products Supplier No. 229046
Sterile, accurate liquid transfer of larger volumes of media and solutionsUniversal fit is compatible with a range of popular pipettesMore precise than serological pipets for volumes of 1 to 10mLTips have molded graduations, with 0.5mL increments