Learn More
US Plastic Corporation 5mL Polycarbonate Vial with 20/415 Closure, each
SDP
Catalog No. NC3355406
Encompass
These 5mL vials are ideal for leakproof shipping of reagents and buffer solutions, for the packaging of serum, veterinary, pharmaceuticals, cosmeceutical, and nutraceutical extracts. Use with most aromatic compounds and essential oils. The 180mL vials can also be used as an outer protective container.Tough polycarbonate vialsPolypropylene leakproof closuresGlass clear allows viewing of contentsReusableAutoclavableDimensions: 0.88" Dia. x 2.05" Hgt. w/out Cap
Catalog No. NC3355406 Supplier US Plastic Corporation Supplier No. 77679
These 5mL vials are ideal for leakproof shipping of reagents and buffer solutions, for the packaging of serum, veterinary, pharmaceuticals, cosmeceutical, and nutraceutical extracts. Use with most aromatic compounds and essential oils. The 180mL vials can also be used as an outer protective container.Tough polycarbonate vialsPolypropylene leakproof closuresGlass clear allows viewing of contentsReusableAutoclavableDimensions: 0.88" Dia. x 2.05" Hgt. w/out Cap