Learn More
MSC CONFORM XT LATEX DISP GLV
Catalog No. 502739224
Encompass
Series TouchNTuff Disposable Gloves: Size Large, 4.7 mil, Uncoated-Coated Latex, Industrial Grade, Unpowdered
Catalog No. 50-273-9224 Supplier MSC Supplier No. 05711023
Series TouchNTuff Disposable Gloves: Size Large, 4.7 mil, Uncoated-Coated Latex, Industrial Grade, Unpowdered