Learn More
Alkali Scientific Qwik-Snap™ 0.5mL MicroTubes, Assorted Colors, 1000/cs
SDP
Catalog No. NC1810217
Encompass_Preferred
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 1,000 0.5mL centrifuge tubes in assorted colors.
Catalog No. NC1810217 Supplier Alkali Scientific Supplier No. C3006A
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 1,000 0.5mL centrifuge tubes in assorted colors.