Learn More
Stellar Scientific 5mL Round Bottom Polystyrene FACS Tubes, 500/case
SDP
Catalog No. NC1731596
Encompass_Preferred
5mL Round Bottom Polystyrene FACS Tubes With 35um Strainer Cap, STERILE, 12x75mm. Each FACS tube is notched near the neck-opening for a perfect fit on multiple FACS equipment including FACS systems from BD Biosciences like the Calibur, Accuri, FACSverse, Aria, Celestia, LSRFortessa, FACSCanto, FACSMelody™ Cell Sorter, and other models.
Catalog No. NC1731596 Supplier Stellar Scientific Supplier No. FSC9005
5mL Round Bottom Polystyrene FACS Tubes With 35um Strainer Cap, STERILE, 12x75mm. Each FACS tube is notched near the neck-opening for a perfect fit on multiple FACS equipment including FACS systems from BD Biosciences like the Calibur, Accuri, FACSverse, Aria, Celestia, LSRFortessa, FACSCanto, FACSMelody™ Cell Sorter, and other models.