Learn More
Sarstedt Inc 5ML SC TUBE 15.3X57 CBS/PK1000
Catalog No. NC9491175 Shop All Sarstedt Inc. Products
Encompass_Preferred
5mL Screw Cap Tube 15.3x57mm Flat Bottom Polypropylene Neutral Cap Enclosed
Catalog No. NC9491175 Supplier Sarstedt Inc Supplier No. 60.558
5mL Screw Cap Tube 15.3x57mm Flat Bottom Polypropylene Neutral Cap Enclosed