Learn More
Alkali Scientific 5ml Screw Cap Tubes, non-sterile, caps packed separately, 5 bags of 100, 500/Pk
SDP
Catalog No. 502336224
Encompass_Preferred
ASI's new 5mL centrifuge tubes take all the conveniences of standard 1.5mL tubes and scales them up for larger volumes. These polypropylene tubes come with an attached cap that not only provides a tight, leak proof seal, it can also be effortlessly opened (or closed) with one hand.In incubation applications, we recommend that you use cap lock clips for incubation and boiling over 80˚C. When the internal pressure approaches an unsafe level, the cap partially opens, softly, in a controlled manner, releasing excess pressure, but preventing any splashing of contents. The center of cap also has a special thin “membrane” area that is designed for easy puncture and access by a syringe/needle. Once punctured, the opening is useful for venting the internal pressure caused by incubation at moderately high temperatures. The tubes are made of maximum clarity, virgin polypropylene and are certified RNase & DNase free. Sterile tubes are also pyrogen free. They are sized to be compatible
Catalog No. 50-233-6224 Supplier Alkali Scientific Supplier No. C2530
ASI's new 5mL centrifuge tubes take all the conveniences of standard 1.5mL tubes and scales them up for larger volumes. These polypropylene tubes come with an attached cap that not only provides a tight, leak proof seal, it can also be effortlessly opened (or closed) with one hand.In incubation applications, we recommend that you use cap lock clips for incubation and boiling over 80˚C. When the internal pressure approaches an unsafe level, the cap partially opens, softly, in a controlled manner, releasing excess pressure, but preventing any splashing of contents. The center of cap also has a special thin “membrane” area that is designed for easy puncture and access by a syringe/needle. Once punctured, the opening is useful for venting the internal pressure caused by incubation at moderately high temperatures. The tubes are made of maximum clarity, virgin polypropylene and are certified RNase & DNase free. Sterile tubes are also pyrogen free. They are sized to be compatible