Learn More
MTC Bio 5mL Screw-cap MacroTube, sterile, w/ screw caps, packed in bags of 100
Catalog No. 502268457
Encompass_Preferred
MTC Bio's 5mL Screw-Cap MacroTubes take all of the conveniences of standard 15mL conical tubes and scales them down for space efficiency and decreased natural resource consumption. Useful from -86°C to 110°C, these tubes are a valuable resource for boiling and for sample preservation at very low temperatures. Choose from gamma sterilized racked packaging with attached caps or non-sterile bulk packaging with separate caps. The foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require about 60% less storage space. Compatible with standard equipment. Centrifuge at up to 25,000xg. RNase, DNase & pyrogen free. Unsurpassed Clarity. Dual graduations at every 0.2mL and 0.5mL
This item is not returnable. View return policy
Catalog No. 50-226-8457 Supplier MTC Bio Supplier No. C2535
MTC Bio's 5mL Screw-Cap MacroTubes take all of the conveniences of standard 15mL conical tubes and scales them down for space efficiency and decreased natural resource consumption. Useful from -86°C to 110°C, these tubes are a valuable resource for boiling and for sample preservation at very low temperatures. Choose from gamma sterilized racked packaging with attached caps or non-sterile bulk packaging with separate caps. The foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require about 60% less storage space. Compatible with standard equipment. Centrifuge at up to 25,000xg. RNase, DNase & pyrogen free. Unsurpassed Clarity. Dual graduations at every 0.2mL and 0.5mL