Learn More
Sarstedt Inc 5ML SCTUBE 15.3X57CBS G/CS1000
Supplier:  Sarstedt Inc 62.558.201
Encompass_Preferred
5ml Screen Printed Screw Cap Tube 15.3x57mm Conical Bottom Skirted Polypropylene Graduations and Writing Block Cap Assembled Sterile
Catalog No. NC9438438