Learn More
Sarstedt Inc 5ML SCTUBE 15.3X57CBS S/PK1000
Catalog No. NC9851632 Shop All Sarstedt Inc. Products
Encompass_Preferred
Tubes, 5mL, 15.3x57mm, Flat Bottom, Polypropylene, Screw Cap, Assembled, Sterile
Catalog No. NC9851632 Supplier Sarstedt Inc Supplier No. 60.558.001
Tubes, 5mL, 15.3x57mm, Flat Bottom, Polypropylene, Screw Cap, Assembled, Sterile