Learn More
Bellco Glass, Inc. 5mL Serological Pipet Individually Wrapped
SDP
Catalog No. 502329448
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identificationPrinted indicator arrow for quickly locating negative graduations when using a pipet controllerPack Description-50 Individually Wrapped/Bag, Case Quantity-200, Sterile-Yes, Non-pyrogenic-Yes, Primary Color-Clear, Secondary Color-Blue, Primary Material-Polystyrene, Maximum Volume-5mL, Graduated-Yes, Graduation Increments (Subdivisions-)1/10mL, RNase/DNase Free-Yes
Catalog No. 50-232-9448 Supplier Bellco Glass, Inc. Supplier No. 229005B00
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identificationPrinted indicator arrow for quickly locating negative graduations when using a pipet controllerPack Description-50 Individually Wrapped/Bag, Case Quantity-200, Sterile-Yes, Non-pyrogenic-Yes, Primary Color-Clear, Secondary Color-Blue, Primary Material-Polystyrene, Maximum Volume-5mL, Graduated-Yes, Graduation Increments (Subdivisions-)1/10mL, RNase/DNase Free-Yes