Learn More
Chemglass Life Sciences Serological Pipette, 5mL, Color Coded Blue, Bulk Packed, Sterile (10 bags of 50)
SDP
Catalog No. 501897705
Encompass_Preferred
Bulk packed serological pipettes are made of super clear clarity polystyrene (PS). Serological pipettes are designed to be used in both tissue culture and bacterial culture experiments, and other liquid handling applications.Edged color-coded band for identificationInclude negative graduations for extra volumeNon-pyrogenic, non-cytotoxic, non-hemolytic, DNase- and RNase-freePolyolefin fiber filter to reduce contaminationMade from high clarity polystyreneSterilized by E-beam
Catalog No. 50-189-7705 Supplier Chemglass Life Sciences Supplier No. CGN3482005
Bulk packed serological pipettes are made of super clear clarity polystyrene (PS). Serological pipettes are designed to be used in both tissue culture and bacterial culture experiments, and other liquid handling applications.Edged color-coded band for identificationInclude negative graduations for extra volumeNon-pyrogenic, non-cytotoxic, non-hemolytic, DNase- and RNase-freePolyolefin fiber filter to reduce contaminationMade from high clarity polystyreneSterilized by E-beam