Learn More
CELLTREAT Scientific Products 5mL Open End Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile pipette
SDP
Catalog No. 50550495
Encompass_Preferred
CELLTREAT Scientific Products 5mL Open End Serological Pipet. Wide, open end is ideal for viscous and high particulate solutions, and slurries such as Sephadex. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in paper/plastic wrappers. 50/bag, 200/case.
Catalog No. 50-550-495 Supplier CELLTREAT Scientific Products Supplier No. 229223B
CELLTREAT Scientific Products 5mL Open End Serological Pipet. Wide, open end is ideal for viscous and high particulate solutions, and slurries such as Sephadex. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in paper/plastic wrappers. 50/bag, 200/case.