Learn More
CELLTREAT Scientific Products 5mL Open End Pipet, Bulk Packed in Bags, Sterile pipette
SDP
Catalog No. 50550496
Encompass_Preferred
CELLTREAT Scientific Products 5mL Serological Pipet, Open end. Wide, open end is ideal for viscous and high particulate solutions, and slurries such as Sephadex. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Bulk packed, Sterile, 25/bag, 500/case.
Catalog No. 50-550-496 Supplier CELLTREAT Scientific Products Supplier No. 229243
CELLTREAT Scientific Products 5mL Serological Pipet, Open end. Wide, open end is ideal for viscous and high particulate solutions, and slurries such as Sephadex. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Bulk packed, Sterile, 25/bag, 500/case.