Learn More
CELLTREAT Scientific Products 5mL Short Serological Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile pipette
SDP
Catalog No. 50202106
Encompass_Preferred
CELLTREAT Scientific Products 5mL Short Serological Pipet. Compact design of short serological pipet provides ergonomic benefit and reduces storage space. Shorter length helps prevent touching pipet tip to surfaces in the hood. Ideal for students and those new to pipetting. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in paper/plastic wrappers. 50/bag, 200/case.
Catalog No. 50-202-106 Supplier CELLTREAT Scientific Products Supplier No. 229206B
CELLTREAT Scientific Products 5mL Short Serological Pipet. Compact design of short serological pipet provides ergonomic benefit and reduces storage space. Shorter length helps prevent touching pipet tip to surfaces in the hood. Ideal for students and those new to pipetting. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in paper/plastic wrappers. 50/bag, 200/case.