Learn More
USA Scientific Inc .5mL SS centrifuge tube,nat
Catalog No. 502745429
Encompass
0.5 mL self-standing screw cap screw cap microcentrifuge tube, natural, with integrated TPE natural color cap, 500/pack
Catalog No. 50-274-5429 Supplier USA Scientific Inc Supplier No. 14059300
0.5 mL self-standing screw cap screw cap microcentrifuge tube, natural, with integrated TPE natural color cap, 500/pack