Learn More
Waters Corp Tip Rack Holder, 5mL, for Andrew+ Pipetting Robot
Catalog No. NC3717982 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 5mL Tip Rack Holder Domino is a machine-readable consumable holder that provides an interface between various consumable types and the Andrew+ pipetting robot. It is designed to hold racks of 50x filtered or non-filtered Sartorius OptiFit 5mL tips. This product is essential for seamless automation of laboratory workflows, featuring QR codes for critical labware information and ARUCO codes for spatial positioning within an Andrew+ automation configuration.
  • Holds racks of 50x filtered or non-filtered Sartorius OptiFit 5mL tips.
  • Creates an interface between consumables and the Andrew+ pipetting robot for laboratory automation.
  • Machine-readable consumable holder.
  • Equipped with QR codes for critical labware information.
  • Equipped with ARUCO codes for spatial positioning.
  • Essential for seamless automation of laboratory workflows.
Catalog No. NC3717982 Supplier Waters Corp Supplier No. 186009599
The 5mL Tip Rack Holder Domino is a machine-readable consumable holder that provides an interface between various consumable types and the Andrew+ pipetting robot. It is designed to hold racks of 50x filtered or non-filtered Sartorius OptiFit 5mL tips. This product is essential for seamless automation of laboratory workflows, featuring QR codes for critical labware information and ARUCO codes for spatial positioning within an Andrew+ automation configuration.
  • Holds racks of 50x filtered or non-filtered Sartorius OptiFit 5mL tips.
  • Creates an interface between consumables and the Andrew+ pipetting robot for laboratory automation.
  • Machine-readable consumable holder.
  • Equipped with QR codes for critical labware information.
  • Equipped with ARUCO codes for spatial positioning.
  • Essential for seamless automation of laboratory workflows.