Learn More
Chemglass Life Sciences Transport Tubes With Double-Threaded Screw Caps, Sterile (Nest)
SDP
Catalog No. 502733396
Encompass_Preferred
Transport tubes designed for secure storage and transport of biological and environmental samples. These self-standing tubes are provided sterile in a nested format and use double-threaded screw caps to ensure reliable sealing during handling and transport.
  • Made from USP VI polypropylene
  • HDPE double-threaded screw cap, separated
  • Electron beam sterilized (SAL = 10⁻⁶)
  • Manufactured in a 100,000-level clean workshop (DNase-free, RNase-free, pyrogen-free, endotoxin-free)
  • Self-standing design for easy handling
  • Temperature range approximately -196°C to 121°C
  • Sealing performance designed to prevent leakage at negative pressure (95 kPa)
  • Supplied in a nested packaging format for efficient processing
Catalog No. 50-273-3396 Supplier Chemglass Life Sciences Supplier No. CGN474235
Add to Cart
Add to Cart
Transport tubes designed for secure storage and transport of biological and environmental samples. These self-standing tubes are provided sterile in a nested format and use double-threaded screw caps to ensure reliable sealing during handling and transport.
  • Made from USP VI polypropylene
  • HDPE double-threaded screw cap, separated
  • Electron beam sterilized (SAL = 10⁻⁶)
  • Manufactured in a 100,000-level clean workshop (DNase-free, RNase-free, pyrogen-free, endotoxin-free)
  • Self-standing design for easy handling
  • Temperature range approximately -196°C to 121°C
  • Sealing performance designed to prevent leakage at negative pressure (95 kPa)
  • Supplied in a nested packaging format for efficient processing