Learn More
Chemglass Life Sciences Chemglass 5 mL transport tubes - sterile, double-threaded screw caps, nest (CGN-4742-45)
SDP
Catalog No. 502733395
Encompass_Preferred
Sterile 5 mL transport tubes with double-threaded screw caps in a nested format, designed for secure sample storage, transport, and freezing. Manufactured from USP VI polypropylene with HDPE caps and electron-beam sterilized for RNase- and pyrogen-free use.
  • Sterile; electron beam sterilized to SAL 10^-6.
  • RNase-free and pyrogen-free.
  • USP VI polypropylene tube with HDPE cap.
  • Self-standing nested format for stable bench handling.
  • Temperature range approximately -196°C to 121°C.
  • Manufactured in a 100,000-level clean workshop.
  • Designed for excellent sealing performance; resists leakage under negative pressure up to 95 kPa.
Catalog No. 50-273-3395 Supplier Chemglass Life Sciences Supplier No. CGN474245
Sterile 5 mL transport tubes with double-threaded screw caps in a nested format, designed for secure sample storage, transport, and freezing. Manufactured from USP VI polypropylene with HDPE caps and electron-beam sterilized for RNase- and pyrogen-free use.
  • Sterile; electron beam sterilized to SAL 10^-6.
  • RNase-free and pyrogen-free.
  • USP VI polypropylene tube with HDPE cap.
  • Self-standing nested format for stable bench handling.
  • Temperature range approximately -196°C to 121°C.
  • Manufactured in a 100,000-level clean workshop.
  • Designed for excellent sealing performance; resists leakage under negative pressure up to 95 kPa.