Learn More
Sarstedt Inc 5ML TUBE 15.3X92FCBS GW/CS1000
Catalog No. NC9785255 Shop All Sarstedt Inc. Products
Encompass_Preferred
5mL Screen Printed Screw Cap Tube 15.3x92mm False Bottom Flat Polypropyelene Graduations and Writing Block Cap Unassembled
Catalog No. NC9785255 Supplier Sarstedt Inc Supplier No. 62.611.009
5mL Screen Printed Screw Cap Tube 15.3x92mm False Bottom Flat Polypropyelene Graduations and Writing Block Cap Unassembled