Learn More
Waters Corp Waters Vial Collection Rack, 12 mm, 24-Vial Capacity, Consumable
Supplier:  Waters Corp 186010531
Encompass_Preferred
This collection rack is a consumable item designed to securely hold 24 vials. It is compatible with the Extraction+ Device and the Collection Rack Domino, and can also be manipulated by the Andrew+ robot when equipped with the Microplate Gripper tool.
  • Holds 24 vials.
  • Compatible with Extraction+ device.
  • Compatible with collection rack domino.
  • Can be moved by Andrew+ robot when using the Microplate Gripper tool.
  • Designed as a consumable rack for ease of use.
Catalog No. 50-226-9417