Learn More
Chemglass Life Sciences Tube, Vacuum Hydrolysis, 4mL Capacity, 10mm Tube OD x 100mm Tube Length
SDP
Catalog No. CG402502
Encompass_Preferred
Tube, Vacuum Hydrolysis, 4mL Capacity, 10mm Tube OD x 100mm Tube Length. Tube is useful in protein hydrolysis, amino acid separations, freeze drying or any application under vacuum.
Catalog No. CG402502 Supplier Chemglass Life Sciences Supplier No. CG402502
Tube, Vacuum Hydrolysis, 4mL Capacity, 10mm Tube OD x 100mm Tube Length. Tube is useful in protein hydrolysis, amino acid separations, freeze drying or any application under vacuum.