Learn More
Thomas Scientific Macrotubes, 5mL, Nonsterile, Green
SDP
Catalog No. 501458817
Encompass_Preferred
MTC Bio’s 5mL MacroTube™ family takes all of the conveniences of 1.5mL tubes and scales them up for larger volumes and throughput applications.  In many cases, they  replace 15mL tubes, dramatically reducing plastic waste, storage space and shipping costs.   Additionally, the 56mm length (vs. 120mm for 15mL tubes) greatly reduces the risk of contamination of pipettor shafts, allowing 1mL or 5mL tips to easily reach the tube bottom. The tubes are made of high clarity, low-binding, virgin polypropylene and are certified RNase and DNase free.  The 16mm diameter is identical to standard 15mL tubes, ensuring compatibility with existing centrifuge rotors.  Other equipment, specially designed for 5mL tubes, is available from Benchmark™ (USA) and German-based equipment manufacturers (eg. dry blocks, tube racks, and  thermal shakers/vortexers). Snap Cap Tubes come with an attached cap that provides a tight, leak-proof seal, and can be effortl
Catalog No. 50-145-8817 Supplier Thomas Scientific Supplier No. 1191K72
MTC Bio’s 5mL MacroTube™ family takes all of the conveniences of 1.5mL tubes and scales them up for larger volumes and throughput applications.  In many cases, they  replace 15mL tubes, dramatically reducing plastic waste, storage space and shipping costs.   Additionally, the 56mm length (vs. 120mm for 15mL tubes) greatly reduces the risk of contamination of pipettor shafts, allowing 1mL or 5mL tips to easily reach the tube bottom. The tubes are made of high clarity, low-binding, virgin polypropylene and are certified RNase and DNase free.  The 16mm diameter is identical to standard 15mL tubes, ensuring compatibility with existing centrifuge rotors.  Other equipment, specially designed for 5mL tubes, is available from Benchmark™ (USA) and German-based equipment manufacturers (eg. dry blocks, tube racks, and  thermal shakers/vortexers). Snap Cap Tubes come with an attached cap that provides a tight, leak-proof seal, and can be effortl