Learn More
Chemglass Life Sciences 5mL Micro Volumetric Flask, Class A, Tolerance: +-0.02mL
SDP
Supplier:  Chemglass Life Sciences CG160304
Encompass_Preferred
5mL Micro Volumetric Flask, Class A, Tolerance: ¿0.02mL. Class ¿A¿ micro volumetric flask calibrated to contain. With 14/20 top outer joint and supplied complete with a CG-3000 14/20 hollow stopper. Flasks have hex base for stability and ¿V¿ shaped interior bottom for total sample retrieval.
Catalog No. CG160304