Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AVIDITY LLC BIO-500 Biotin Solution, 5 mM d-biotin, 10 mL, 10 mM Bicine buffer, pH 8.3, Filter sterilized (0.22 μm), Room temperature shipping, -20 °C storage

Catalog No. NC9854810
Encompass

The AVIDITY LLC BIO500 is a 5mM d-biotin solution supplied in a 10mL volume, designed for critical molecular biology applications. It is particularly suitable for in-vivo biotinylation culture inductions utilizing pBirAcm BirA over-expression plasmids and AviTag™-protein fusion vectors. The solution is also appropriate for large-scale BirA reactions requiring supplemental d-biotin and for supplementing induction growth media. Filter sterilized through a 0.22μm filter, this product ensures sterility for sensitive scientific protocols.

  • Concentration: 5 mM d-biotin
  • Volume: 10 mL
  • Buffer: 10 mM Bicine
  • pH: 8.3
  • Sterility: Filter Sterilized (0.22 μm)
  • Shipping Condition: Room temperature
  • Storage Temperature: -20 °C
  • Primary Application: In-vivo biotinylation
  • Secondary Application: Large scale BirA reactions

Catalog No. NC9854810 Supplier AVIDITY LLC Supplier No. BIO500
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The AVIDITY LLC BIO500 is a 5mM d-biotin solution supplied in a 10mL volume, designed for critical molecular biology applications. It is particularly suitable for in-vivo biotinylation culture inductions utilizing pBirAcm BirA over-expression plasmids and AviTag™-protein fusion vectors. The solution is also appropriate for large-scale BirA reactions requiring supplemental d-biotin and for supplementing induction growth media. Filter sterilized through a 0.22μm filter, this product ensures sterility for sensitive scientific protocols.

  • Concentration: 5 mM d-biotin
  • Volume: 10 mL
  • Buffer: 10 mM Bicine
  • pH: 8.3
  • Sterility: Filter Sterilized (0.22 μm)
  • Shipping Condition: Room temperature
  • Storage Temperature: -20 °C
  • Primary Application: In-vivo biotinylation
  • Secondary Application: Large scale BirA reactions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.