Learn More
New Era Enterprises 5MM BODY, 3MM STEM NMR TUBES
Catalog No. NC1870120
Encompass_Preferred
5mm Body, 3mm Stem NMR Tubes for reduced volume
Catalog No. NC1870120 Supplier New Era Enterprises Supplier No. NEH5/3BR
5mm Body, 3mm Stem NMR Tubes for reduced volume