Learn More
Electron Microscopy Sciences Round Cover Slip German Glass #1.5, 5 mm
SDP
Catalog No. 50949439
Encompass_Preferred
These coverglass which are made from German glass can be used for growing and culturing cells that normally have poor adhesion to plastic or regular glass surfaces. Available with #1.5 (0.16 – 0.19 mm) thickness. 5mm fits inside the 96 well plates.
Diameter: 5 mm
Catalog No. 50-949-439 Supplier Electron Microscopy Sciences Supplier No. 7229605
These coverglass which are made from German glass can be used for growing and culturing cells that normally have poor adhesion to plastic or regular glass surfaces. Available with #1.5 (0.16 – 0.19 mm) thickness. 5mm fits inside the 96 well plates.
Diameter: 5 mm