Learn More
Perkin Elmer US LLC Cell Adapter for Fluorescence Micro Cells - Qty. 4
Catalog No. 509052842
Encompass_Preferred
The Cell Adapter for Fluorescence Micro Cells allows 5 mm path length micro cells to be used in any 10 mm path length cuvette accessory. Qty. 4.
Catalog No. 50-905-2842 Supplier Perkin Elmer US LLC Supplier No. L2250139
The Cell Adapter for Fluorescence Micro Cells allows 5 mm path length micro cells to be used in any 10 mm path length cuvette accessory. Qty. 4.