Learn More
Wilmad Labglass 5mm Economy Tube PTFE Plug, 100/Pk
Catalog No. NC1240005
Encompass
These PTFE NMR/EPR Tube Caps are machined to exact specifications with smaller gyroradius and more homogeneous mass distribution than disposable caps providing better spinning stability. They are recommended for experiments at high - ultra high field and those using Chloroform as the reference solution. They will only fit our Precision tubes due to the tight tolerance.
Catalog No. NC1240005 Supplier Wilmad Labglass Supplier No. WG1265100
These PTFE NMR/EPR Tube Caps are machined to exact specifications with smaller gyroradius and more homogeneous mass distribution than disposable caps providing better spinning stability. They are recommended for experiments at high - ultra high field and those using Chloroform as the reference solution. They will only fit our Precision tubes due to the tight tolerance.