Learn More
Azzota Corp 5mm Pathlength Standard Cuvettes - 1.7Ml Pathlength - 5mm Inside Width - 10mm Volume - 1.7Ml Dimension - 45X12.5X7.5mm With Cover And Round Bottom Optical Glass (340Nm-2500Nm)
Catalog No. 507532885
Encompass_Preferred
5mm Pathlength Standard Cuvettes - 1.7ml Pathlength - 5mm Inside width - 10mm Volume - 1.7ml Dimension - 45x12.5x7.5mm With cover and round bottom Optical Glass (340nm-2500nm)
Catalog No. 50-753-2885 Supplier Azzota Corp Supplier No. G103
5mm Pathlength Standard Cuvettes - 1.7ml Pathlength - 5mm Inside width - 10mm Volume - 1.7ml Dimension - 45x12.5x7.5mm With cover and round bottom Optical Glass (340nm-2500nm)