Learn More
Ops Diagnostics 5 mm Grinding Balls, 440C Stainless Steel, Pack of 2500
Catalog No. NC1419150
Encompass
OPS Diagnostics' 5mm Stainless steel grinding balls can be used with 4 ml and 15 ml vials to homogenize. 5mm grinding balls are made of 440C stainless steel which is slightly magnetic, this allows them to be retrieved with a magnet while making them resistant to corrosion. Grinding balls are treated to remove residual oils and contaminants prior to packaging. The grinding balls are ready-to-use. These 5 mm grinding balls are suitable for use with the Qiagen Tissuelyser bead dispenser.
Catalog No. NC1419150 Supplier Ops Diagnostics Supplier No. GBSS 196250010
OPS Diagnostics' 5mm Stainless steel grinding balls can be used with 4 ml and 15 ml vials to homogenize. 5mm grinding balls are made of 440C stainless steel which is slightly magnetic, this allows them to be retrieved with a magnet while making them resistant to corrosion. Grinding balls are treated to remove residual oils and contaminants prior to packaging. The grinding balls are ready-to-use. These 5 mm grinding balls are suitable for use with the Qiagen Tissuelyser bead dispenser.