Learn More
Ace Glass, Inc. 5MM ID GRADED SEALS 5MM ID GRADED SEALS
SDP
Supplier:  Ace Glass, Inc. 845506
Encompass
5mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-591