Learn More
Norell Inc 5mm OD, 178mm LONG, RATED UP TO 700MHz, SAMPLE VAULT SERIES NMR TUBES FOR AUTOMATED SAMPLE TUBE SYSTEM, 96PK, INCLUDES CLOSED PORT SAMPLE VAULT CAPS
SDP
Catalog No. NC0765498
Encompass_Preferred
5mm OD, 178mm LONG, RATED UP TO 700MHz, SAMPLE VAULT SERIES NMR TUBES FOR AUTOMATED SAMPLE TUBE SYSTEM, 96PK, INCLUDES CLOSED PORT SAMPLE VAULT CAPS
Catalog No. NC0765498 Supplier Norell Inc Supplier No. SVCP517896PK
5mm OD, 178mm LONG, RATED UP TO 700MHz, SAMPLE VAULT SERIES NMR TUBES FOR AUTOMATED SAMPLE TUBE SYSTEM, 96PK, INCLUDES CLOSED PORT SAMPLE VAULT CAPS