Learn More
Azzota Corp 5mm Path Fluorometer 1.7ml Qz
Catalog No. 5000887149
Encompass_Preferred
45x12 5x12 5mm 5mm Pathlength Standard Fluorometer Cuvettes 1 7ml Quartz 190nm-2500nm Four polished windows
Catalog No. 50-008-87149 Supplier Azzota Corp Supplier No. Q203
45x12 5x12 5mm 5mm Pathlength Standard Fluorometer Cuvettes 1 7ml Quartz 190nm-2500nm Four polished windows