Learn More
Starna Cells Inc 5MM PATH LENGTH STD QUARTZ CEL
SDP
Catalog No. NC9981025
Encompass_Preferred
Quartz, Spectrosil, Path 5mm L, 12.5W x 7.5L x 45mm H Exterior, 10.5W x 5mm L Interior, 1.700mL Nominal Volume
Catalog No. NC9981025 Supplier Starna Cells Inc Supplier No. 1Q5
Quartz, Spectrosil, Path 5mm L, 12.5W x 7.5L x 45mm H Exterior, 10.5W x 5mm L Interior, 1.700mL Nominal Volume