Learn More
Azzota Corp 5mm Pathlength Standard Cuvettes - 1.7Ml Pathlength - 5mm Inside Width - 10mm Volume - 1.7Ml Dimension - 45X12.5X7.5mm With Cover And Round Bottom Quartz (190Nm-2500Nm)
Catalog No. 507532891
Encompass_Preferred
5mm Pathlength Standard Cuvettes - 1.7ml Pathlength - 5mm Inside width - 10mm Volume - 1.7ml Dimension - 45x12.5x7.5mm With cover and round bottom Quartz (190nm-2500nm)
Catalog No. 50-753-2891 Supplier Azzota Corp Supplier No. Q103
5mm Pathlength Standard Cuvettes - 1.7ml Pathlength - 5mm Inside width - 10mm Volume - 1.7ml Dimension - 45x12.5x7.5mm With cover and round bottom Quartz (190nm-2500nm)