Learn More
Starna Cells Inc 5MM UV CELL FAR UV QUARTZ
SDP
Catalog No. NC1619075
Encompass_Preferred
Spectrosil ® Far UV Quartz windows, Useable range: 170 to 2700 nm, 5mm cell
Catalog No. NC1619075 Supplier Starna Cells Inc Supplier No. 9BQ5
Spectrosil ® Far UV Quartz windows, Useable range: 170 to 2700 nm, 5mm cell