Learn More
Medchemexpress LLC 5MPN | 47208-82-2 | MFCD00089445 | 99.9% | 305.33 | C15H19N3O4 | 10MG
SDP
Catalog No. 5000239132
Encompass_Preferred
5MPN is a selective, orally active inhibitor of 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 4 (PFKFB4), used as a research compound to probe tumor sugar metabolism and inhibit cancer cell proliferation in vitro.
  • Selective PFKFB4 inhibition, competitive at the F6P binding site (Ki = 8.6 μM).
  • Targets tumor sugar metabolism and suppresses proliferation of multiple human cancer cell lines.
  • High purity (~99.9%) suitable for biochemical assays.
  • Available in small research quantities such as 10 mg and as 10 mM DMSO solution.
  • Molecular weight 305.33 and molecular formula C15H19N3O4 for analytical reference.
Catalog No. 50-002-39132 Supplier Medchemexpress LLC Supplier No. HY12398110MG
5MPN is a selective, orally active inhibitor of 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 4 (PFKFB4), used as a research compound to probe tumor sugar metabolism and inhibit cancer cell proliferation in vitro.
  • Selective PFKFB4 inhibition, competitive at the F6P binding site (Ki = 8.6 μM).
  • Targets tumor sugar metabolism and suppresses proliferation of multiple human cancer cell lines.
  • High purity (~99.9%) suitable for biochemical assays.
  • Available in small research quantities such as 10 mg and as 10 mM DMSO solution.
  • Molecular weight 305.33 and molecular formula C15H19N3O4 for analytical reference.