Learn More
Medchemexpress LLC HY-123981 5mg , 5MPN CAS:47208-82-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1239815MG
Encompass_Preferred
HY-123981 5mg 5MPN CAS: 47208-82-2 5MPN is a first-in-class, potent, orally active and selective 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 4 (PFKFB4) inhibitor. 5MPN appears to be a competitive inhibitor of the F6P binding site (Ki=8.6 μM). 5MPN does not inhibit PFK-1 or PFKFB3. 5MPN targets the sugar metabolism of tumors and suppresses proliferation of multiple human cancer cell lines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1710