Learn More
Cayman Chemical 5mthyltetrahydrofolc ACD 10mg
SDP
Catalog No. 502971825
Encompass_Preferred
A biologically active form of folic acid that functions, in conjunction with vitamin B12, as a methyl-group donor involved in the conversion of homocysteine to methionine
Catalog No. 50-297-1825 Supplier Cayman Chemical Supplier No. 1615910 MG
A biologically active form of folic acid that functions, in conjunction with vitamin B12, as a methyl-group donor involved in the conversion of homocysteine to methionine