Learn More
Trajan Scientific 5M X 0.1MM ID - BPX5 0.1UM GC COLUMN
SDP
Catalog No. 502725261
Encompass_Preferred
5M X 0.1MM ID - BPX5 0.1UM GC COLUMN
Catalog No. 50-272-5261 Supplier Trajan Scientific Supplier No. 0540990
Add to Cart
Add to Cart
5M X 0.1MM ID - BPX5 0.1UM GC COLUMN