Learn More
Cayman Chemical 5'-N-Ethylcarboxamidoadenosine (hydrate), Purity: ≥98%, 5 mg
SDP
Catalog No. 502994312
Encompass_Preferred
An adenosine receptor agonist (Kis = 1,880, 6,660, and 3.5 nM for human adenosine subtypes A1, A2A, and A3, respectively); potent vasodilator that can inhibit platelet aggregation by increasing cAMP (EC50 = 3.1 µM)
Catalog No. 50-299-4312 Supplier Cayman Chemical Supplier No. 214205 MG
An adenosine receptor agonist (Kis = 1,880, 6,660, and 3.5 nM for human adenosine subtypes A1, A2A, and A3, respectively); potent vasodilator that can inhibit platelet aggregation by increasing cAMP (EC50 = 3.1 µM)