Learn More
Cayman Chemical 5′-O-(4,4′-Dimethoxytrityl)thymidine, Purity: ≥98%, CAS: 40615-39-2, 50 mg
SDP
Catalog No. 502978863
Encompass_Preferred
A derivative of thymidine and synthetic precursor; has been used as a precursor in the synthesis of oligo-2’-deoxyribonucleotides and phosphoramidate derivatives
Catalog No. 50-297-8863 Supplier Cayman Chemical Supplier No. 3080250 MG
A derivative of thymidine and synthetic precursor; has been used as a precursor in the synthesis of oligo-2’-deoxyribonucleotides and phosphoramidate derivatives