Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 5P Urine Reagent Strips for BLO/pH/PRO/KET/GLU, CLIA Waived

Catalog No. 23111420 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Strips/Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-420 100 Strips/Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-420

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the semi-quantitative detection of the following analytes in urine: Glucose, Ketone, pH, Blood, Protein. Professional, in vitro diagnostic use. A substantial equivalent to the leading national brand. Use with the Fisherbrand Urine Analyzer or read visually.

A CLIA Waived, easy-to-use system that provides fast, highly-accurate results for the general evaluation of health, aiding in the diagnosis and monitoring of metabolic or systemic diseases that affect kidney function, endocrine disorders and diseases or disorders of the urinary tract.

Features:

  • Analytical sensitivity comparable to the market leader
  • High quality color chart ensures accurate visual reading
  • 2 year shelf life for unopened canisters
  • 3 month shelf life for strips in opened canisters

Specifications

CE Marker No
Clia Complexity Waived
Detectable Analytes Glucose, Protein, Ketone, pH, Blood
Format Urine Reagent Strip
Sample Type Urine
Sensitivity Substantial equivalent to the leading national brand
Type Urine Reagent Strip
Disposable Single-use
Quantity 100 Strips/Bottle
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.